Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "prepared-media-in-bottles"

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Pepton water buffered, Filling: 225 ml, Size: 250 ml, Standard: -, Application: Enrichment, non selective
Supplier:  VWR Chemicals
Description:   S.O.C. Medium is used in the final step of bacterial cell transformation to obtain maximal transformation efficiency of E. coli. S.O.C. LB brot...
Catalogue Number: (1B1493-1L)
Supplier:  VWR Chemicals
Local Article Number:: VWRC1B1493-1L
Description:   A complete medium for the propagation and maintenance of yeast. The composition of YPD is (w/v) 1% Yeast Extract, 2% Peptone, and 2% D-Glucose.
UOM:  1 * 1 l
Supplier:  Hach
Description:   Selectiv for Total Coliform Bacteria and Escherichia coli. Broth in pre-filled bottles.
Catalogue Number: (516-9079)
Supplier:  Pall Laboratory
Local Article Number:: PALL4313
Description:   M-Endo broth is used for the estimation of coliform bacteria in water samples using the membrane filtration technique. It is possible to remove bacter...
UOM:  1 * 1 items
Catalogue Number: (MILXSTBMRFA34)
Supplier:  MILLIPORE
Local Article Number:: MILXSTBMRFA34
Description:   For sterility testing (fi ltration and direct inoculation) and bioburden testing. In the screw cap version, the rimless cap design minimizes the risks...
UOM:  1 * 4 items
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 6  of 6