Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "blotting-reagents"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/49/58/18944958.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"43"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Antibody Signal Enhancer is a Western blotting antibody diluent that increases intensity of protein detection up to 5-fold compared to conventional bl...
Supplier:  VWR Chemicals
Description:   VisiGlo™ HRP Chemiluminescent Substrates are luminol-based chemiluminescent substrates designed for rapid and sensitive detection of peroxidase labell...
Supplier:  VWR Chemicals
Description:   DAB is commonly used for detection of horseradish peroxidase in membrane-bound and in situ applications.
Supplier:  VWR Chemicals
Local Article Number:: VWRCJ348-50T
Description:   OPD tablets are widely used to detect horseradish peroxidase activity in ELISA and solution assays. Absorbs at 492 nm. Each tablet contains 15 mg of O...
UOM:  1 * 50 Tablet
Catalogue Number: (E885-200ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCE885-200ML
Description:   Complete substrate system includes 20 DAB substrate tablets, peroxidase solution and a mixing/dispensing dropper bottle for fast and easy preparation....
UOM:  1 * 200 ml
Supplier:  VWR Chemicals
Local Article Number:: VWRCN656-KIT
Description:   UnDo™ X-Ray Film Background Reducer kit easily and proportionally reduces the signal on overexposed film. The kit includes two solutions that are used...
UOM:  1 * 1 KIT
Supplier:  GE Healthcare - Life Sciences
Local Article Number:: GEHENAMP100
Description:   Amplify™ is a fast and convenient fluorographic reagent for ³⁵S-, ¹⁴C- and ³H-labelled samples in gels. Amplify™ can increase sensitivity of detection...
UOM:  1 * 1 l
Catalogue Number: (RPN5781)
Supplier:  GE Healthcare - Life Sciences
Local Article Number:: GEHERPN5781
Description:   ECF™ Western blotting reagent packs use alkaline phosphatase conjugated antibodies to catalyse the formation of stable fluorophores that emit light wh...
UOM:  1 * 1 items
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 43
  1  2  3  4  5  6  Next