Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Molecular Weight Markers"

https://au.vwr.com/stibo/smallweb/std.lang.all/95/65/18989565.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"34"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (1B1491-25RXN)
Supplier:  VWR Chemicals
Description:   RNA ladder, RiboReady™ Color Micro
UOM:  1 * 1 KIT
Supplier:  VWR Chemicals
Description:   Protein MW markers, Prism
Catalogue Number: (1B1617-500UL)
Supplier:  VWR Chemicals
Description:   Protein marker, low range, BLUE DUO™
UOM:  1 * 500 µl
Catalogue Number: (1B1492-25RXN)
Supplier:  VWR Chemicals
Description:   RNA ladder, 1 kb, RiboReady™ Color
UOM:  1 * 1 KIT
Catalogue Number: (1B1584-500UL)
Supplier:  VWR Chemicals
Description:   Protein marker, high range, BLUE DUO™
UOM:  1 * 1 KIT
Catalogue Number: (1B1527-500UL)
Supplier:  VWR Chemicals
Description:   Protein MW marker, mid-range, Blue Duo™
UOM:  1 * 500 µl
Supplier:  VWR Chemicals
Description:   Protein MW markers, BlueStep™ low range, pre-stained
UOM:  1 * 0,5 ml
Catalogue Number: (J383-200UL)
Supplier:  VWR Chemicals
Description:   Protein MW markers, Precise™
UOM:  1 * 200 µl
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 34
  1  2  3  4  5  Next