Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Electrophoresis Reagents"

https://au.vwr.com/stibo/smallweb/std.lang.all/51/84/18945184.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
 
 
SearchResultCount:"51"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (J449-200UL)
Supplier:  VWR Chemicals
Description:   High range protein molecular weight marker offers 5 bands ranging from 40,0 - 212,0 kDa. The 200 μl size provides sufficient material for 40 assa...
UOM:  1 * 200 µl
Catalogue Number: (J450-200UL)
Supplier:  VWR Chemicals
Description:   Mid/low range protein molecular weight marker offers 7 bands ranging from 14,4 - 97,4 kDa. The 200 μl size provides sufficient material for 40 as...
UOM:  1 * 200 µl
Catalogue Number: (J383-200UL)
Supplier:  VWR Chemicals
Description:   Precise™ protein molecular weight marker offers 7 bands ranging from 15,0 - 150,0 kDa. There is sufficient material for 40 assays.
UOM:  1 * 200 µl
Supplier:  VWR Chemicals
Description:   BlueStep™ protein molecular weight marker, broad range, pre-stained has 9 bands ranging from 9,0 to 200,0 kDa. Pre-stained bands are easily transferre...
UOM:  1 * 0,5 ml
Supplier:  VWR Chemicals
Description:   BlueStep™ protein molecular weight marker, low range, pre-stained has 6 bands ranging from 19,0 to 120,0 kDa. Pre-stained bands are easily transferred...
UOM:  1 * 0,5 ml
Catalogue Number: (E255-150UG)
Supplier:  VWR Chemicals
Description:   High-Range DNA MW (molecular weight) marker offers 10 fragments ranging from 1503 to 48 502 bp. There is sufficient material for 75 - 150 assays.
UOM:  1 * 150 µG
Catalogue Number: (E261-100UG)
Supplier:  VWR Chemicals
Description:   Low-Range I™ marker offers 14 fragments ranging from 224 to 14 140 bp. Combine 5 μl of marker with 5 μl of 2X loading buffer and load into well. There...
UOM:  1 * 100 µG
SDS
Catalogue Number: (E854-100RXN)
Supplier:  VWR Chemicals
Description:   PCR DNA Marker™ offers 8 fragments ranging from 50 to 2000 bp. The marker contains high intensity reference bands that are ideal for quick sizing of P...
UOM:  1 * 1 KIT
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 51
  1  2  3  4  5  6  7  Next