Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    VWR Chemicals (60) GE Healthcare - Life Sciences (88) Thermo Scientific (26)

  • Search Within Results

You Searched for: "PCR Reagents"

https://au.vwr.com/stibo/smallweb/std.lang.all/96/20/18989620.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"58"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Taq DNA Polymerase Master Mix
Supplier:  VWR Chemicals
Description:   Taq DNA Polymerase
Catalogue Number: (1B1326-50RXN)
Supplier:  VWR Chemicals
Description:   Ready One-Step RT-PCR Kit
UOM:  1 * 1 KIT
Supplier:  VWR Chemicals
Description:   dNTP
SDS
Supplier:  VWR Chemicals
Description:   Ready PCR Mix, 2X
Catalogue Number: (733-1361)
Supplier:  VWR Chemicals
Description:   Betaine Enhancer, 5 M (5X)
UOM:  1 * 5 ml
Supplier:  VWR Chemicals
Description:   Hot Start Taq PCR Mastermix, 2X
Supplier:  VWR Chemicals
Description:   Extender™ Taq DNA Polymerase Blend, 500 U
UOM:  1 * 1 KIT
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 58
  1  2  3  4  5  6  7  8  Next