Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

You Searched for: "Electrophoresis Buffers"

https://au.vwr.com/stibo/smallweb/std.lang.all/93/52/18989352.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
 
 
SearchResultCount:"156"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   PBS and TBS Wash buffers
SDS
Supplier:  VWR Chemicals
Description:   Stripping buffer, Gentle ReView™
SDS
Catalogue Number: (M245-10.4G-5PK)
Supplier:  VWR Chemicals
Description:   Wash buffer, PBS with 0,05% Tween® 20, pH 7,4
UOM:  1 * 5 PAC
SDS
Supplier:  VWR Chemicals
Description:   TRIS-glycine-SDS buffer, solid for 10X concentrated solution
SDS
Supplier:  VWR Chemicals
Description:   Laemmli loading buffer, 4X
SDS
Supplier:  VWR Chemicals
Description:   TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer) Electran® for electrophoresis
UOM:  1 * 5 l
SDS
Supplier:  VWR Chemicals
Description:   TBE (Tris-Borate-EDTA) buffers
SDS
Supplier:  VWR Chemicals
Description:   Tris-Acetate-EDTA (TAE) buffer
SDS
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 156
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next