Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "hazard-warning-labels"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/53/13/18945313.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Collection
Description:   Labels aid in general workplace safety and efficiency by quickly and simply identifying contaminated equipment containers, rooms, or materials. A bioh...
Supplier:  VWR Products
Description:   These labels, made of PE film and with the GHS symbols, meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures. The...
Catalogue Number: (116-1095)
Supplier:  Bel-Art
Local Article Number:: BELAF133590000
Description:   Stick-on dots with the radiation symbol on yellow background, which enables easy labelling of radioactive vessels.
UOM:  1 * 1 Roll
Supplier:  Merck
Local Article Number:: MERC1.00801.0001
Description:   With implementation of CLP/EU-GHS (Global Harmonised System) directive in January 2009, the procedures for the classification, labelling, and packagin...
UOM:  1 * 1 items
Supplier:  PRECISION DYNAMICS CORP.-TIMEM
Description:   Hazard Warning Label, Radioactive material, yellow, 76,2×19,05 mm
Supplier:  PRECISION DYNAMICS CORP.-TIMEM
Description:   Self-adhesive labels with Biohazard pictogram.
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 6  of 6