Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "aprons"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/49/07/18944907.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"20"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Products
Description:   This disposable apron is made from a mixture of 65% LDPE and 35% LLDPE.
Supplier:  Bel-Art
Description:   These aprons made of vinyl have a stitched cotton binding securing all edges.
Catalogue Number: (BELAF246020000)
Supplier:  Bel-Art
Local Article Number:: BELAF246020000
Description:   This lightweight, yet effective Beta-blocking neoprene apron blocks up to 96% of the high energy beta rays from Strontium-90 and over 98% of the beta ...
UOM:  1 * 1 items
Catalogue Number: (BURK2507-1001)
Supplier:  BÜRKLE
Local Article Number:: BURK2507-1001
Description:   Plastic apron made of PVC that is suitable for industrial applications.
UOM:  1 * 1 items
Supplier:  TEMPSHIELD INC
Description:   Cryo-Aprons® are multi-layer insulated Aprons designed to provide protection to the torso and legs from contact with ultra cold surfaces in cryogenic ...
Catalogue Number: (113-0479)
Supplier:  DuPont
Local Article Number:: DUPOPA30SWHL0
Description:   Specially designed for use with Tyvek® apparel, this apron can help offer enhanced protection for body parts that are more exposed to hazardous substa...
UOM:  1 * 100 items
Supplier:  DuPont
Description:   Specially designed for use with Tychem® apparel, these aprons can help offer enhanced protection for body parts that are more exposed to hazardous sub...
Catalogue Number: (DALL4101180)
Supplier:  HONEYWELL SAFETY
Local Article Number:: DALL4101180
Description:   These aprons made of grey and blue split leather (heat retro-reflective) have been designed to protect welders against molten metal splashes.
UOM:  1 * 1 items
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 20
  1  2  3  Next