Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "deep-well-microplates"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/95/44/18709544.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"17"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Collection
Description:   PP. These microplates are uniform dimensionally so that all covers, whether round or square, fit interchangeably on other round or square well micropl...
Supplier:  VWR Products
Description:   PP. These deep well plates are designed for robotic use. They have smooth inside surfaces to ensure that whole sample can be pipetted.
Supplier:  VWR Products
Description:   These PP deep well plates feature double alignment corner notches and are labelled on three sides with an alphanumeric grid.
Supplier:  VWR Products
Description:   PP. These plates hold a full 2,5 ml (2,2 ml with sealing mat in place) and work as storage plates or reservoirs.
Supplier:  EPPENDORF
Description:   PP. For processing, transport and storage of liquids. The well geometry enables high-speed pipetting, excellent mixing and high-speed centrifugation t...
Supplier:  Corning Life Science
Description:   PP, non treated, without lid. Standard and large volume 96-well microplates and storage blocks to meet assay and storage requirements.
Supplier:  Corning Life Science
Description:   PP. Standard and large volume 384-well microplates and storage blocks to meet assay and storage requirements.
Supplier:  Corning Life Science
Description:   PVC, clear, non sterile, without lid. Untreated microplates for solution-based assays, serial dilutions, and general storage applications.
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 17
  1  2  3  Next