Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "electroporation-electrodes"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/93/53/18989353.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"16"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  BTX (GENETRONICS INC.)
Description:   Electrode, Electroporation, platinum, Tweezertrodes™
Supplier:  BTX (GENETRONICS INC.)
Description:   This electrode is designed for delicate and/or difficult tissue transfection. Ex vivo electropoartion is an efficient, effective method to introduce g...
Supplier:  BTX (GENETRONICS INC.)
Description:   AgilePulse™ needle array electrodes are miniature needles designed to provide superior, highly uniform electric fields in dermal or muscular tissue as...
Catalogue Number: (BTXI45-0510)
Supplier:  BTX (GENETRONICS INC.)
Description:   This needle style platinum electrode is specifically designed for in vivo applications on the most fragile of tissue types, such as brain tissue. The ...
UOM:  1 * 1 KIT
Supplier:  BTX (GENETRONICS INC.)
Description:   This tissue chamber is specifically designed to handle ex-vivo tissue samples that are either larger than normal or have a unique shape making it diff...
Supplier:  BTX (GENETRONICS INC.)
Description:   Genetrodes™ are paired, reusable, needle style electrodes that are ideal for in vivo and in ovo electroporation applications, including ...
Catalogue Number: (732-0025)
Supplier:  BTX (GENETRONICS INC.)
Description:   The Petri Pulser™ electrode is a reusable electroporation applicator designed to fit into each single well of a 6-well plate or an individual 35 mm di...
UOM:  1 * 1 items
Supplier:  BTX (GENETRONICS INC.)
Description:   2-Needle Array™ electrodes are needle-style electrodes specifically designed for in vivo drug or gene delivery applications. The electrode cons...
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 16
  1  2  Next