Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "cell-spreaders"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/50/09/18945009.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"12"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Products
Description:   PP, sterile (individually wrapped) or non sterile (bulk packed). Convenient and economical, these cell spreaders are designed to simplify bacterial cu...
Supplier:  VWR Products
Description:   PS, sterile. Designed for spreading and dispersing liquids across the surface of agar culture plates. Used for performing bacterial (for example,...
Supplier:  VWR Products
Description:   These PS, sterile disposable L-shaped spreaders enable even spreading of liquid samples across the surface of agar plates without gouging or cutting t...
Supplier:  VWR Products
Description:   These PS, sterile disposable T-shaped spreaders are designed for spreading and dispersing liquids across the surface of agar culture plates. Their des...
Supplier:  VWR Products
Description:   PS, sterile. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of li...
Catalogue Number: (89042-018)
Supplier:  VWR Collection
Local Article Number:: VWRU89042-018
Description:   PP, sterile, disposable L-shaped cell spreaders are ideal for spreading bacterial cultures evenly.
UOM:  1 * 500 items
Catalogue Number: (HEATHS8151)
Supplier:  HEATHROW
Local Article Number:: HEATHS8151
Description:   Triangular shaped spreading bar has an edgeless design which prevents tearing of the agar and head shape that distributes pressure evenly for smooth s...
UOM:  1 * 500 items
Catalogue Number: (HEATHS86655)
Supplier:  HEATHROW
Local Article Number:: HEATHS86655
Description:   Stainless steel spreader for use with 90 mm plates. Smooth triangular end makes spreading easy - the upturned end will not cut into agar.
UOM:  1 * 1 items
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 12
  1  2  Next