Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "presaturated-wipes"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (115-0128)
Supplier:  VWR Products
Local Article Number:: VWRI115-0128
Description:   These non woven wipes are constructed of cellulose and polyester and moistened with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reage...
UOM:  1 * 100 items
SDS
Supplier:  MICRONOVA
Local Article Number:: MICRSSW6-01IR
Description:   SilSat wipes address the problem of using lubricant in a controlled application by offering high grade silicone on a pre-saturated wipe. Each polyeste...
UOM:  1 * 24 items
Catalogue Number: (115-0301)
Supplier:  Texwipe
Local Article Number:: TEXWTX1051
Description:   PolySat® is constructed from a 100% melt-blown polypropylene substrate that is pre-wetted with 70% isopropyl alcohol and packaged in a resealable Flex...
UOM:  1 * 1.200 items
Supplier:  MICRONOVA
Local Article Number:: MICRMMR-01
Description:   These biodegradbale poly blend wipes are pre-moistened with a moisturizing lotion. They are gentle enough for the face, but can also be used to remove...
UOM:  1 * 250 items
Supplier:  CONTEC
Local Article Number:: CONTSWSIL0006
Description:   Proper lubrication is essential to the efficient operation of most equipment and many processes. SATWipes® Silicone Lubricating wipes provide this lub...
UOM:  1 * 750 items
Catalogue Number: (MMMA504)
Supplier:  3M
Local Article Number:: MMMA504
Description:   The 3M™ respirator cleaning wipes 504 clean and refresh elastomeric facepiece respirators, eliminating dirt and perspiration.
UOM:  1 * 1 items
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 6  of 6