Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "disposable-wipes-and-towels"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/51/31/18945131.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
 
 
SearchResultCount:"36"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Products
Description:   These wet-laid, double re-creped, general purpose wipes, made from wood pulp with binder, can be used for many different appplications.
Supplier:  VWR Products
Description:   These spunlace wipes, made from 55% wood pulp and 45% polyester, are extremely strong, even when wet.
Catalogue Number: (115-0128)
Supplier:  VWR Products
Local Article Number:: VWRI115-0128
Description:   These non woven wipes are constructed of cellulose and polyester and moistened with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reage...
UOM:  1 * 100 items
SDS
Supplier:  VWR Products
Description:   Absorbent paper, suitable for cleaning and drying of glass containers, tubes, microscopic plates, bottles, etc.
Supplier:  VWR Products
Description:   These bleached spunlace wipes made from 55% wood pulp and 45% polyester are extremely strong, even when wet.
Supplier:  VWR Products
Description:   These meltblown wipes, made from 100% PP, have a very high absorption capacity.
Supplier:  VWR Products
Description:   Toalhetes de velo absorventes, macios, espessos e com baixo nível de fibras. Para utilização em várias tarefas no laboratório, indústria electrónica e...
Supplier:  VWR Products
Description:   These very soft wipes, made from a spunlace fabric, consisting of 60% viscose and 40% polyester, are ideal for sensitive surfaces and light-duty clean...
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 36
  1  2  3  4  5  Next