Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "reagent-reservoirs"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/97/15/18989715.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"10"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR LabAdvantage
Description:   These reagent reservoirs made of PS or PVC are designed to accommodate all types of pipettors from single channel to 16-channel.
Supplier:  VWR Products
Description:   Designed for use in multi-channel pipetting applications, these disposable reservoirs accommodate all types of pipettes up to 12-channel width.
Supplier:  Corning Life Science
Description:   White PS, sterile, disposable.
Supplier:  Bel-Art
Description:   PS, white. These plastic pipetting reservoirs have a 100 ml capacity for extended multi channel pipetting into microwell plates, Mini-Tubes, Cube 2ube...
Supplier:  HEATHROW
Description:   Clear PVC or white PS, disposable.
Catalogue Number: (BELAF378801200)
Supplier:  Bel-Art
Local Article Number:: BELAF378801200
Description:   PP. This chemical resistant 12 channel reagent reservoir can be used with single, 8 and 12 channel pipettes.
UOM:  1 * 8 items
Catalogue Number: (HEATHS20821A)
Supplier:  HEATHROW
Local Article Number:: HEATHS20821A
Description:   PP. This non sterile solution basin/reagent reservoir has a standard 50 ml basin on one side and 12 individual 5 ml basins on the other side.
UOM:  1 * 25 items
Supplier:  Corning Life Science
Description:   White PS, sterile, disposable.
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 10
  1  2  Next