Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "cell-culture-media"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/53/55/18945355.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Corning Life Science
Description:   Used as part of an integrated system designed to create in vitro intestinal models.
UOM:  1 * 1 KIT
Catalogue Number: (734-2385)
Supplier:  Corning Life Science
Description:   Endothelial culture medium is optimised to promote rapid growth (3- to 5-fold increase in cell number over five days) of endothelial cells from a vari...
UOM:  1 * 1 KIT
Catalogue Number: (734-2386)
Supplier:  Corning Life Science
Description:   Hepatocyte culture medium is a serum-free, fully-defined medium optimised for hepatocyte differentiation.
UOM:  1 * 500 ml
Catalogue Number: (ICNA091440049)
Supplier:  MP Biomedicals
Description:   Ham's nutrient mixtures were developed for the clonal growth of several chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells.
UOM:  1 * 100 ml
Supplier:  MP Biomedicals
Description:   Minimum Essential Medium (MEM) is a general use medium ideal for the growth and maintenance of a wide range of mammalian cell types. MEM with Earle's ...
Supplier:  MP Biomedicals
Description:   Basal Medium Eagle is suited for the isolation and cultivation of non-fastidious and fastidious microorganisms from a variety of clinical and non-clin...
UOM:  1 * 10 l
Supplier:  MP Biomedicals
Description:   Murashige and Skoog Medium is widely used plant tissue culture growth medium. It has proven effective in the culture of tissue derived from moncotyled...
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 7  of 7