Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "ionic-strength-adjustment-buffers"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/40/31/18374031.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Local Article Number:: VWRC160847G
Description:   Total ionic strength adjustment buffer solution for use with selective ion electrodes.
UOM:  1 * 5 l
SDS
Supplier:  Hanna
Local Article Number:: HANNHI7023L
Description:   TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations
UOM:  1 * 500 ml
Supplier:  METTLER / TOLEDO
Local Article Number:: METT51350106
Description:   TISAB 3 SOLUTION, 250ML 1 * 250 ml
UOM:  1 * 250 ml
Supplier:  WTW
Description:   Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluo...
Catalogue Number: (1.16770.0500)
Supplier:  Merck
Local Article Number:: MERC1.16770.0500
Description:   TISAB III solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations
UOM:  1 * 500 ml
SDS
Catalogue Number: (1.15368.0500)
Supplier:  Merck
Local Article Number:: MERC1.15368.0500
Description:   TISAB SOLUTION FOR FLUORIDE DETERMINATION 1 * 500 ml
UOM:  1 * 500 ml
SDS
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 6  of 6