Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "running-buffers"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"47"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (E526-500ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCE526-500ML
Description:   MOPS is a zwitterionic buffer used as a running buffer for denaturing agarose gel electrophoresis of RNA. Having a buffering range from 6.5 - 7.9, MOP...
UOM:  1 * 500 ml
Catalogue Number: (K855-500ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCK855-500ML
Description:   Useful for high resolution of proteins on neutral pH SDS-PAGE gels.
UOM:  1 * 500 ml
Supplier:  VWR Chemicals
Description:   Liquid concentrate used for protein electrophoresis to maintain pH and temperature of the environment as well as provide ions that carry electrical cu...
SDS
Supplier:  VWR Chemicals
Description:   TBE is optimised for DNA electrophoresis techniques using both acrylamide and agarose. Buffers are available as liquid concentrate or Ready-Pack™ (a f...
SDS
Catalogue Number: (1B1373-500ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRC1B1373-500ML
Description:   RNA Gel running buffer, 10X is the direct replacement for MOPS buffer, which is typically used in denaturing formaldehyde RNA gels.
UOM:  1 * 500 ml
Supplier:  VWR Chemicals
Description:   TAE is an extensively used buffer for agarose gel electrophoresis applications requiring high resolution and separation of high molecular weight, doub...
SDS
Supplier:  VWR Chemicals
Local Article Number:: VWRCM149-40L
Description:   The 40L pack size contains sufficient dry powder to prepare 40 litres of 1X buffer. 1X solution contains: 0.025 M Tris, 0.192 M glycine, and 0.1% SDS.
UOM:  1 * 40 l
SDS
Supplier:  VWR Chemicals
Local Article Number:: VWRCE442-500ML
Description:   Electrophoresis, Running Buffer, TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
UOM:  1 * 500 ml
SDS
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 47
  1  2  3  4  5  6  Next