Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "loading-buffers"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"28"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (N717-2X1.5ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCN717-2X1.5ML
Description:   RNA EZ-Vision® is a non mutagenic fluorescent dye for staining RNA bands resolved on denaturing agarose gels containing formaldehyde. Supplied in a 1,...
UOM:  1 * 1 Each
Catalogue Number: (M260-5.0ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCM260-5.0ML
Description:   The NEXT GEL® Sample Loading Buffer is recommended for sample denaturation and loading with the NEXT GEL® product line.
UOM:  1 * 5 ml
Supplier:  VWR Chemicals
Description:   RNA loading buffers (2X) are premixed loading dyes for the preparation of RNA samples for agarose or polyacrylamide electrophoresis.
Supplier:  VWR Chemicals
Description:   LDS Sample Loading Buffer, Reducing (4X) is used to prepare protein samples for denaturing polyacrylamide gel electrophoresis using Bis-Tris and Laemm...
Supplier:  VWR Chemicals
Description:   LDS Sample Loading Buffer, Non-reducing (4X) is used to prepare protein samples for denaturing polyacrylamide gel electrophoresis using Bis-Tris and L...
Catalogue Number: (E270-5ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCE270-5ML
Description:   Protein gel loading buffer, 2X is used for denaturing protein electrophoresis.
UOM:  1 * 5 ml
Supplier:  VWR Chemicals
Description:   Laemmli loading buffer, 4X is formulated for use in Laemmli SDS-PAGE, a widely used system for protein electrophoresis. Mixing Laemmli loading buffer,...
SDS
Supplier:  Alfa Aesar
Description:   Electrophoresis
SDS
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 28
  1  2  3  4  Next