Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "lysis-buffers"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/40/31/18374031.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"31"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (N653-100ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCN653-100ML
Description:   Lysis buffer for cultured mammalian cells containing SDS and deoxycholate for solubilisation of proteins from cytoplasm, membranes, and nuclei. Compat...
UOM:  1 * 100 ml
Catalogue Number: (J619-500ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCJ619-500ML
Description:   NP-40 Lysis Buffer is manufactured from Nonidet P-40 Substitute.
UOM:  1 * 500 ml
Catalogue Number: (M334-100ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCM334-100ML
Description:   Detergent-free buffer for isolation of proteins from tissue culture cells. Cell lysis and the subsequent isolation of the crude cytoplasmic fraction c...
UOM:  1 * 100 ml
Supplier:  Alfa Aesar
Description:   E. coli lysis buffer 1 * 500 ml
SDS
Supplier:  Alfa Aesar
Description:   Liquid
SDS
Supplier:  Alfa Aesar
Description:   RBC lysis buffer for human 1 * 250 ml
SDS
Supplier:  Alfa Aesar
Description:   MES lysis buffer with Triton|r X-100 1 * 500 ml
SDS
Catalogue Number: (J62809.AK)
Supplier:  Alfa Aesar
Local Article Number:: ALFAJ62809.AK
Description:   HEPES lysis buffer with Triton|r X-100 (2X) 1 * 250 ml
UOM:  1 * 250 ml
SDS
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 31
  1  2  3  4  Next