Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "hybridisation-buffers"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (E717-50ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRCE717-50ML
Description:   A blocking reagent used to help prevent probe binding to open membrane space and non target nucleic acids.
UOM:  1 * 50 ml
SDS
Catalogue Number: (0973-50ML)
Supplier:  VWR Chemicals
Local Article Number:: VWRC0973-50ML
Description:   Hybridization buffers offer blocking and hybridization for optimized complimentary base pairing of DNA and RNA in Southern blot, Northern blot, dot bl...
UOM:  1 * 50 ml
Supplier:  VWR Chemicals
Description:   SSC buffer is a hybridisation buffer for Northern and Southern blots.
SDS
Supplier:  VWR Chemicals
Local Article Number:: VWRCE257-50ML
Description:   A blocking reagent for Northern and Southern blots that prevents probe binding on open membrane space and non target nucleic acids.
UOM:  1 * 50 ml
SDS
Supplier:  Alfa Aesar
Description:   Electrophoresis, Hybridisation Buffers
SDS
Supplier:  Alfa Aesar
Description:   Electrophoresis, Hybridisation Buffers
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   A major limitation of membrane hybridisation is speed. At commonly used probe concentrations (2 to 10 ng/ml), hybridisation normally takes 16 hours to...
Supplier:  Alfa Aesar
Description:   SSPE (20X) 1 * 1 l
SDS
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 9
  1  2  Next