Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "safety-caps-and-safety-waste-caps"

https://au-bot.vwr.com/stibo/smallweb/std.lang.all/97/57/18989757.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"16"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Products
Description:   Extract the solvents or mobile phases safely from reservoirs and bottles, without being exposed to hazardous gases and solvent vapours. High quality m...
Supplier:  VWR Products
Description:   Collect your solvent waste safely without being exposed to solvent vapours or hazardous gases. High quality materials (PTFE, PP) ensure maximum safety...
Catalogue Number: (590-1521)
Supplier:  VWR Products
Local Article Number:: VWRI590-1521
Description:   Safety caps with empty level state display ensure maximum safety for laboratory workers and environment.
Extract the solvent safely from the 5 l,...
UOM:  1 * 1 items
Supplier:  Merck
Description:   Specially developed screw caps for direct instrument connection of Merck S 40 threaded bottles ensure safe handling of solvents in the lab and protect...
Supplier:  SCAT Europe
Description:   These pure PTFE and PE-HD SafetyWasteCaps with connector for exhaust filter are resistant against organic solvents, acids and lyes.
Supplier:  SCAT Europe
Description:   For safe disposal of fluid waste. Collect your solvent waste safely, without being exposed to hazardous gases and solvent vapours. The integrated exha...
Supplier:  SCAT Europe
Description:   Fire resistant SCAT caps incorporate a self-extinguishing component with a special plastic. In the event of a fire, the SCAT fire resistant material b...
Supplier:  SCAT Europe
Description:   Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. With integrated air valve for p...
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 16
  1  2  Next