Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "prepared-media-in-ampoules"

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"11"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Pall Laboratory
Description:   A wide variety of sterile ampoule media for common microbiological analyses, in clear glass or plastic ampoules. Supplied pre-measured for maximum eff...
Supplier:  Hach
Description:   Microbiological media in ampoules, m-TGE, For: Total Count
Catalogue Number: (HACH2428350)
Supplier:  Hach
Local Article Number:: HACH2428350
Description:   Microbiological media in ampoules, m-Green yeast and mold, 2 ml plastic ampule, For: Yeast and Moulds
UOM:  1 * 50 items
Supplier:  GE Healthcare - Whatman
Local Article Number:: WHAT10496124
Description:   Nutrient media ( 2 ml ampoules) specifically suited to the cultivation of colonies after microfiltration.
UOM:  1 * 50 items
Supplier:  Hach
Description:   Selectivity: fecal coliform bacteria.
Supplier:  Hach
Description:   m-Endo broth detects total coliform bacteria (colonies with a greenish-gold sheen). For potable water samples, confirm with Lauryl Tryptose and Brilli...
Catalogue Number: (HACH2428520)
Supplier:  Hach
Local Article Number:: HACH2428520
Description:   Selectivity: Fecal Coliform Bacteria. For drinking water, wastewater, ambient water, food, and beverage applications
UOM:  1 * 20 items
Supplier:  Hach
Description:   In 24 hours, Hach's m-ColiBlue24® broth give the simultaneous results for total coliforms and E. coli. Hach's ready-to-use media eliminates measuring,...
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 11
  1  2  Next