Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "general-purpose-digital-thermometers"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"34"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Products
Description:   These thermometers contain red organic liquid against white back glass and are ideal for use in chemical laboratories, universities, scientific instit...
Supplier:  VWR Products
Description:   Large-digit triple-display thermometer, Traceable®, 99×111×22 mm, 113 g
Supplier:  VWR Products
Description:   These thermometers fit into flasks, cuvettes, test tubes and beakers for temperatures reading in food, gases, samples, reagents, water baths, ovens, i...
Supplier:  VWR Products
Description:   Digital dial thermometer, VWR® Traceable® Ultra™
Supplier:  VWR Products
Description:   Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens.
Catalogue Number: (620-1933)
Supplier:  VWR Products
Local Article Number:: VWRI620-1933
Description:   This ready to use thermometer with removeable probe features automatic or manual choice of the measuring range. It can be used for general purpose app...
UOM:  1 * 1 items
Supplier:  VWR Products
Description:   Triple display thermometer, Traceable®
Supplier:  VWR Products
Description:   These digital thermometers measure the temperature of wet or dry environments including freezers, refrigerators, water baths, aquariums and incubators...
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 8  of 34
  1  2  3  4  5  Next