Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "environmentally-friendly-glass-thermometers"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Products
Description:   These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. The numbers are print...
Supplier:  VWR Products
Description:   These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chem...
Supplier:  VWR Products
Description:   These thermometers are made with a 100% non toxic, non hazardous, biodegradable liquid. Easy to read, thanks to a high contrast green against white ba...
Supplier:  VWR Products
Description:   Each easy-to-read thermometer includes an individualised thermometer calibration report stating corrections to one-tenth of the smallest division, uni...
Supplier:  VWR Products
Description:   Thermometers with Enviro-Safe® liquid are equipped with a closed metal case which protects from breakage. Ideal for field studies, waste water treatme...
Supplier:  Bel-Art
Local Article Number:: BELAB605020000
Description:   General purpose liquid in glass thermometer, Enviro-Safe®, −35…+50 °C, Length: 300 mm, Organic
UOM:  1 * 1 items
Price on Request
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1300 727 696.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, please contact our customer service team on 1300 727 696.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service at 1300 727 696.
1 - 6  of 6