Would you like to log in again?
VWR LabAdvantage Pipette Tips
Savings!
VWR LabAdvantage Pipette Tips
VALID UNTIL: 30/06/2017
Special sales pricing on 10, 200 and 1000μL tips
Standard tips for general pipetting applications. Designed to fit most research-grade pipettors.
  • Available with low binding surface technology for reduced sample retention
  • Certified free from RNases, DNases, human DNA, PCR inhibitors and pyrogens
  • Standardization tips are ideal for multi-channel pipettors: Thin collar requires less force for loading and ejecting tips
  • XT tips (extended length): Ideal for pipetting from deep vessels
  • OneTouch tips create a leak proof seal: Easy to seal, easy to eject
Description Pack Size Best.-Nr. Promo Price Quantity
Standard Tip, 100-1000μL, blue, sterile 1 * 1,000 items 613-3033AU
Standard Tip, 100-1000μL, natural, bulk 1 * 1,000 items 613-3034AU
Standard Tip, 100-1000μL, natural, sterile, low bind 1 * 1,000 items 613-3036AU
One Touch Tip, 1-330μL, yellow 1 * 960 items 613-3040AU
One Touch Tip, 1-330μL, yellow, sterile 1 * 960 items 613-3041AU
One Touch Tip, 50-1250μL, blue, sterile 1 * 768 items 613-3044AU
One Touch Tip, 1-20μL, red, sterile 1 * 960 items 613-3046AU
One Touch Tip, 1-330μL, yellow, sterile 1 * 960 items 613-3048AU
One Touch Tip, 1-330μL, yellow, sterile 1 * 960 items 613-3049AU
One Touch Tip, 50-1250μL, blue, sterile 1 * 768 items 613-3050AU
One Touch Tip, 50-1250μL, blue, sterile 1 * 768 items 613-3051AU
Ultra G, with graduation marks, 0.1-10µL, natural, bulk 1 * 1,000 items 613-3065AU
One Touch Tip, 1-200μL, yellow, bulk 1 * 1,000 items 613-3057AU
Standard Tip, 100-1000μL, blue, bulk 1 * 1,000 items 613-3058AU