Would you like to log in again?
Pyrex Quickfit Range On Sale
Save 40%!
Pyrex Quickfit Range On Sale
VALID UNTIL: 30/06/2017
Huge discounts on a huge range!
Quickfit pioneered the concept of interchangeable joints in 1934. Today, the Quickfit name is renowned throughout the world for quality and reliability. Quickfit ground-glass joints are precision manufactured with an accurate taper and smooth finish for leak-free performance and assured interchangeability. The Quickfit range comprises over 1,000 products, enabling the user to construct the widest choice of assemblies. Quickfit glassware is manufactured exclusively from Pyrex borosilicate glass. This type of glass has a very low co-efficient of expansion, allowing use at both elevated and sub-zero temperatures. Quickfit is the first choice of scientists who demand maximum performance and value for money.
  • Unbadged ground glass sockets
    • Free from any ceramic markings
    • Avoids the problem of the badge charring at high temperatures which can give an untidy appearance
  • High quality joints manufactured to an accuracy which surpasses the requirements of ISO 383
  • Accurate taper and fine surface finish reduce possibility of joint 'seizure' or leakage
  • With round profile bead on top of socket for extra mechanical strength
Description UOM Promo Code Promo Price Quantity
Condenser Liebig Borosilicate 14/23 Cone 1 * 1 items 201-8270AU
Condenser Liebig Borosilicate 19/26 Cone 1 * 1 items 201-8271AU
Condenser Liebig Borosilicate 24/29 Cone 1 * 1 items 201-8272AU
Condenser Liebig Borosilicate 24/29 Cone 1 * 1 items 201-8274AU
Adapter Reduction Cone 19/26 Socket 14/23 1 * 2 items 201-2357AU
Adapter Reduction Cone 24/29 socket 14/23 1 * 2 items 201-0255AU
Adapter Reduction Cone 24/29 socket 19/26 1 * 2 items 201-0111AU
Adapter Reduction Cone 29/32 socket 24/29 1 * 2 items 201-8349AU
Flask 5ml NS14/23 Pear Shaped 23x67mm 1 * 1 items 212-0600AU
Flask 50mL NS14/23 2 Necks Pear Shape 115mm 1 * 1 items 212-0602AU
Flask Round Bottom Borosilicate 10mL 1 * 5 items 201-8466AU
Flask Round Bottom Borosilicate 10mL 1 * 5 items 201-8467AU
Flask 10mL R Bottom 14/23 Socket Septum 1 * 1 items 212-0604AU
Flask Round Bottom Quickfit 100mL 1 * 5 items 201-8468AU
Flask Round Bottom 100mL Socket 19/26 1 * 5 items 201-8469AU
Flask Round Bottom 100mL 1 * 1 items 201-8470AU
Flask Round Bottom 100mL NS 24/29 1 * 5 items 201-8471AU
Flask Round Bottom 100mL 1 * 1 items 201-8472AU
Flask 2 Neck Round Bottom 100mL 1 * 1 items 201-0563AU
Flask Round Bottom 2 Neck 100mL 1 * 1 items 201-0608AU
Flask Round Bottom Quickfit 150mL 1 * 5 items 201-8479AU
Flask Round Bottom Quickfit 150mL 1 * 5 items 201-8481AU
Flask Round Bottom Borosilicate 1L 1 * 2 items 201-8484AU
Flask Round Bottom 2 Neck 1L 1 * 1 items 201-8486AU
Flask Round Bottom Borosilicate 25ml 1 * 5 items 201-8497AU
Flask Round Bottom Short Neck 25ml 19/16 1 * 5 items 201-8498AU
Flask Round Bottom Short Neck 25ml 24/29 1 * 5 items 201-8499AU
Flask Short Neck Round Bottom 250mL 1 * 5 items 201-8500AU
Flask Round Bottom 250mL Socket 24/29 1 * 5 items 201-8503AU
Flask Round Bottom 250mL 1 * 1 items 201-0712AU
Flask 250mL Round Bottom 2 Necks 24/29 Cone 1 * 1 items 201-0713AU
Flask Round Bottom 3 Necks 250mL 1 * 1 items 201-8506AU
Flask Round Bottom 3 Necks 250mL 1 * 5 items 201-8509AU
Flask Round Bottom Short Neck 250mL 1 * 5 items 201-8510AU
Flask Round Bottom 3 Necks 2l 1 * 1 items 201-8515AU
Flask Round Bottom Short Neck 5ml 1 * 5 items 201-8528AU
Flask Round Bottom Short Neck 5ml 1 * 5 items 201-8529AU
Flask Round Bottom Short Neck 50mL 1 * 5 items 201-8530AU
Flask Round Bottom Short Neck 50mL 1 * 5 items 201-8531AU
Flask Round Bottom Short Neck 50mL 1 * 5 items 201-8533AU
Flask Round Bottom Short Neck 500mL 1 * 5 items 201-8534AU
Flask Round Bottom Medium Neck 500mL 1 * 2 items 201-8535AU
Flask Round Bottom Short Neck 500mL 1 * 5 items 201-8536AU
Flask Round Bottom 3 Necks 500mL 1 * 1 items 201-8537AU
Flask Volumetric Class A W/Stopper 5ml 1 * 5 items 612-0442AU
Flask Volumetric Class A W/Stopper 20mL 1 * 5 items 612-0444AU
Flask Volumetric 100mL Class A 1 * 5 items 612-3667AU
Flask Volumetric Class A Plastic 250mL 1 * 2 items 612-0447AU
Clip Polyacetal Joint 14/23 Yellow 1 * 10 items 201-8566AU
Clip Polyacetal Joint 19/26 Blue 1 * 10 items 201-8567AU
Clip Polyacetal Joint 24/29 Green 1 * 10 items 201-8568AU
Clip Polyacetal Joint 29/32 Red 1 * 10 items 201-8569AU
Adapter Tubing Right Angle 14/23 Cone 1 * 1 items 201-0567AU
Adapter Tubing Right Angle 24/29 Cone 1 * 1 items 201-0568AU
Adapter Stopcock with Cone 14/23 1 * 1 items 201-0263AU
Adapter Stopcock with Cone 19/26 1 * 1 items 201-8596AU
Adapter Stopcock with Cone 24/29 1 * 1 items 201-8599AU
Adapter Tubing Borosilicate Straight 19/26 socket 1 * 1 items 201-0894AU
Tube Air/Steam Inlet Straight Cone 14/23 1 * 1 items 201-3056AU
Test Tube Borosilicate 54ml 150mm 24/29 Socket 1 * 5 items 201-8626AU
Connector Screw Thread 130mm 24/29 Cone 1 * 1 items 201-0774AU
Connector Screw Thread 215mm 24/29 Cone 1 * 1 items 201-0776AU
Thermometer -10 to +250 Hg Fill 14/23 1 * 1 items 620-1962AU
Adapter Receiver Borosilicate 110mm Height, 14/23 1 * 1 items 271-0013AU
Stirrer Shaft Anchor Blade 450mm 1 * 1 items 441-0361AU
Washer Felt 6mm Spare for Stirrer Gland 1 * 10 items 201-8733AU
Adapter Straight Cone 14/23 Screw thread 1 * 2 items 201-0219AU
Adaptor for Screw Thread 7-8.5mm Cap 1 * 2 items 201-0696AU
Adaptor for Screw Thread 6-7mm Cap 1 * 2 items 201-0220AU
Adaptor for Screw Thread 7-8.5mm Cap 1 * 2 items 201-0225AU
Adaptor for Screw Thread 6-7mm Cap 1 * 2 items 201-0697AU
Adaptor for Screw Thread 10.5-11.5mm Cap 1 * 2 items 201-0699AU
Adapter Expansion 10/19 Sock 14/23 Cone 1 * 2 items 201-0704AU
Adapter Expansion 19/26 Sock 14/23 Cone 1 * 2 items 201-0229AU
Adapter Expansion 24/29 Sock 14/23 Cone 1 * 2 items 201-0230AU
Adapter Expansion 24/29 Sock 19/26 Cone 1 * 2 items 201-0235AU
Adapter Expansion 29/32 Sock 19/26 Cone 1 * 2 items 201-0237AU
Adapter Expansion Sock 29/32 Cone 24/29 1 * 2 items 201-0238AU
Adapter Expansion Sock 34/35 Cone 24/29 1 * 2 items 201-0706AU