Would you like to log in again?
Falcon Serological Pipettes
Limited time only!
Falcon Serological Pipettes
VALID UNTIL: 30/06/2017
28% off the range of Falcon pipettes
Falcon® Serological pipettes
  • Polystyrene, transparent, graduated and sterile
  • Coloured top with cotton plug and coloured packaging for easy identification
  • Negative graduations for extra capacity and reverse
    graduations (except 1 ml)
  • Non pyrogenic
Packaging: Individually wrapped in paper-peelable bags or bulk packaged in bags.
Description Pack size Cat. No. Promo price Quantity
Serological Pipet, 1mL, 0.01mL graduations, Sterile, 25 per bag 1 * 1,000 items 734-0335AU
Serological Pipet, 2mL, 0.01mL graduations, Sterile, 100 per box 1 * 1,000 items 734-0336AU
Serological Pipet, 2mL, 0.01mL graduations, Sterile, 25 per bag 1 * 1,000 items 734-0337AU
Serological Pipet, 25mL, 0.25mL graduations, Sterile, 20 per bag 1 * 200 items 734-0338AU
Serological Pipet, 1mL, 0.01mL graduations, Sterile, 100 per box 1 * 1,000 items 734-0340AU
Serological Pipet, 25mL, 0.25mL graduations, Sterile, 50 per bag 1 * 200 items 734-0343AU
Serological Pipet, 5mL, 0.1mL graduations, Sterile, 25 per bag 1 * 500 items 734-0344AU
Serological Pipet, 10mL, 0.1mL graduations, Sterile, 25 per bag 1 * 500 items 734-0345AU
Serological Pipet, 25mL, 0.5mL graduations, Extended, 50 per bag 1 * 200 items 734-0347AU
Serological Pipet, 5mL, 0.1mL graduations, Individually Packed, 50 per bag 1 * 200 items 734-0350AU
Serological Pipet, 50mL, 1mL graduations, Sterile, 25 per bag 1 * 100 items 734-0351AU
Serological Pipet, 10mL, 0.1mL graduations, Sterile, 50 per bag 1 * 200 items 734-0352AU
Serological Pipet, 100mL, 1mL graduations, Sterile, 5 per bag 1 * 50 items 734-0355AU