Would you like to log in again?
Azlon Beakers
Great for sampling and storage!
Azlon Beakers
VALID UNTIL: 30/06/2017
Specials on Azlon beakers
Azlon® Low Form beakers
Polypropylene, translucent beakers
Ideal for general laboratory use. With spout and exceptionally easy to read blue graduations. Straight sided.
  • Good chemical resistance
  • Resistant to heat up to +135 °C
  • Smooth surfaces for quick, easy cleaning
ISO 7056
Description Pack Size Promo Code Promo Price Quantity
Beaker, Low Form, Polypropylene, Graduated, 10mL 1 * 10 items 213-0269AU
Beaker, Low Form, Polypropylene, Graduated, 25mL 1 * 10 items 213-0058AU
Beaker, Low Form, Polypropylene, Graduated, 50mL 1 * 10 items 213-0060AU
Beaker, Low Form, Polypropylene, Graduated, 100mL 1 * 10 items 213-0062AU
Beaker, Low Form, Polypropylene, Graduated, 250mL 1 * 10 items 213-0064AU
Beaker, Low Form, Polypropylene, Graduated, 400mL 1 * 10 items 213-0065AU
Beaker, Low Form, Polypropylene, Graduated, 500mL 1 * 10 items 213-0067AU
Beaker, Low Form, Polypropylene, Graduated, 600mL 1 * 5 items 213-0068AU
Beaker, Low Form, Polypropylene, Graduated, 1000mL 1 * 5 items 213-0070AU
Beaker, Low Form, Polypropylene, Graduated, 2000mL 1 * 1 items 213-0072AU