Would you like to log in again?
IKA HS7 Hotplate Stirrer Promotion
New low price!
IKA HS7 Hotplate Stirrer Promotion
VALID UNTIL: 31/03/2017
The IKA HS7 has a magnetic stirrer and ceramic heating plate, which offers excellent chemical resistance
  • Powerful motor for stirring quantities of up to 10 L (H2O)
  • Fixed safety circuit of 550°C
  •  Hot Top indicator >> hot surface warning to prevent burns!
  •  Exact temperature setting via digital display (LED)
  •  Bushing according to DIN 12878 for connecting a contact thermometer, e.g. ETS-D5, enables precise temperature control
  •  Digital error code display
  •  Elevated control panel for protection against leaking liquids
* Register your device online with IKA and receive a free 12 month warranty extension – click here for details
Description Promo Code Promo Price Quantity
IKA Hotplate Stirrer HS7 442-1103AU