Would you like to log in again?
Elga Flex Water System Promotion
25% OFF
Elga Flex Water System Promotion
VALID UNTIL: 31/03/2017
The award winning PURELAB Flex is an adaptable single system for all your pure and ultrapure needs
Now only $7,999 (ex GST), down from $10,780 (ex GST)
This state-of-the-art system integrates in-hand monitoring and accurate pure water dispensing in one unique product. This makes general to highly specialised applications in chemistry and life sciences research and testing laboratories effortless.
The flexible hand-held dispenser has an easy-to-read digital display, which shows both volume and water purity (either resistivity or conductivity and Total Organic Carbon) giving users complete confidence in the water quality, allowing them to visibly monitor the water purity as they are dispensing.
The dispense handset has precise fingertip control (like a pipette) and is intuitive to use, therefore minimal training is required. The handset can also be easily programmed with pre-set volumes, which can be repeated with the single press of a button.
Flexible dispensing by:
  • Variable flow - drop by drop or up to 2 litres per minute
  • Auto-volume dispense from 50ml to 60 litres and repeat dispensing
  • Hands free with optional foot pedal
  • Locked dispense for glassware filling
  • Intuitive to use, ergonomic handset desig
  • Clear water purity display for absolute confidence as you dispense
  • Real-time TOC monitoring for critical analytical applications
  • Prioritized information shown at all times (system status, TOC, alarm)
  • Point of use filters for multiple applications
  • Quick and easy sanitization
Description Promo Code Promo price Quantity
PURELAB Flex 3 - Type 1 Water System (from potable tap water source) 171-0872AU
PURELAB Flex 3 - Type 1 Water System (from potable tap water source) 171-0873AU