Would you like to log in again?
Total Organic Carbon (TOC)
Total Organic Carbon (TOC)
Total Organic Carbon (TOC) is the amount of carbon bound in an organic compound and is often used as a non-specific indicator of water quality. The rapid method of TOC determination by colorimetric method makes this an easy method to ensure great results.
  • Rapid digestion
  • Choice of photometers and spectrophotometers and ready-to-use TOC tubes and powder pillows.